Disposable PES Bottle Top Filters
Actual product may vary
Fisherbrand
Disposable PES Bottle Top Filters
Increase sample protection with Fisherbrand Sterile Disposable Bottle Top Filters with PES Membranes. The polyethersulfone (PES) membranes have low extractables and low protein binding for use in sterile filtration and clarification of cell culture media and serum.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Pore SizeDiameter (Metric) Membrane
FISFB12566510500 mL0.20 μm75 mm
FISFB12566509150 mL0.45 μm50 mm
FISFB125665121000 mL0.20 μm91 mm
Catalog number FISFB12566510
Price (SGD)
195.00
Case
Volume (Metric):
500 mL
Pore Size:
0.20 μm
Diameter (Metric) Membrane:
75 mm
Price (SGD)
195.00
Case
  • PES membrane is low protein binding to maintain protein balance
  • PES membrane has low extractables to maintain media purity
  • Marked with catalog number, membrane type, pore size, lot number and expiration date for easy traceability
  • Non-pyrogenic and non-cytotoxic
  • Gamma radiation-sterilized and individually bagged for a 5-year sterile shelf life
  • Bottom screw threads for attachment to bottles with 45 mm neck sizes
  • 0.2μm units for sterile filtration
  • 0.45μm units for fluid clarification and particle removal
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceNon-pyrogenic per USP Standard Lot-to-Lot
DisposableSingle-use
Material (Membrane)PES
Pore Size0.20 μm
SterilitySterile
For Use With (Application)Sterile Filtration of media and serum
TypeBottle Top Unit
Volume (Metric)500 mL
Diameter (Metric) Membrane75 mm
Unit SizeCase