Bench Protector
Bench Protector
Actual product may vary
Bench Protector
Bench Protector
Bench Protector
Bench Protector
Bench Protector
Fisherbrand™ Bench Protector features absorbent paper to capture spills, backed with an impermeable polyethylene to protect the covered surface from contamination
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W)Description
FISFB120073020.92 x 50 mRoll
FISFB1200730046 x 57 cmSheet
FISFB12007301Roll
Catalog number FISFB12007302
Price (SGD)
301.00
EA
Add to cart
Dimensions (L x W):
0.92 x 50 m
Description:
Roll
Price (SGD)
301.00
EA
Add to cart
Features:
  • Absorbent layer of pure cellulose with smooth, firm surface
  • One side coated with impermeable polyethylene
  • Dry and liquid samples can be recovered
  • Can be incinerated for disposal
Specifications
Product TypeBench Protector
MaterialCellulose/Polyethylene
Absorbancy25 mg/cm2
Length (English)164 ft.
Length (Metric)50 m
Width (English)18.11 in.
Width (Metric)46 cm
Dimensions (L x W)0.92 x 50 m
ColorWhite
DescriptionRoll
DisposableSingle-use
For Use With (Application)Protects benchtops, floors and carts with this highly absorbent cover
Quantity1
Unit SizeEA