Check Order Status
Electroporation Cuvettes Plus™
Actual product may vary
Electroporation Cuvettes Plus™
Disposable, color-coded cuvets fit most electroporators. Sterilized by gamma irradiation and supplied individually wrapped.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
Width (Metric) Gap
FISFB104
800 μL
4 mm
FISFB102
400 μL
2 mm
2 Options
Catalog number
FISFB104
Price (SGD)
374.00
PK
Add to cart
Capacity (Metric):
800 μL
Width (Metric) Gap:
4 mm
Price (SGD)
374.00
PK
Add to cart
Disposable, color-coded cuvets are compatible with most electroporators.
Clear, medical grade polycarbonate for high visibility
Polished aluminum lowers arcing frequency
Round cap allows single-handed removal
FB102 has imbedded aluminum electrodes
Specifications
Sterility
Sterile
Type
Electroporation Cuvet
Capacity (Metric)
800 μL
Material
Polycarbonate
Includes
Imbedded aluminum electrodes
Width (Metric) Gap
4 mm
For Use With (Equipment)
Electroporator