Petri Dishes Specialty
Petri Dishes Specialty
Actual product may vary
Petri Dishes Specialty
Multiple sizes and styles for variety of applications
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeSize
FISFB0875711ZDeep Dish100 x 20mm
FISFB0875711ADish100 x 15 mm
FISFB0875711YZMicroplate35 x 10mm
FISFB0875711CContact Plate65 x 15mm
Catalog number FISFB0875711Z
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Type:
Deep Dish
Size:
100 x 20mm
Multiple sizes and styles for variety of applications
  • All dishes manufactured from virgin medical grade polystyrene
  • All dishes sterilized using e-beam sterilization
  • Deep style petri dishes available - designed for increased media volumes, allowing for longer culture times
  • Contact plates available for collection in environments where contact is required to collect the sample
  • Convenient packaging in easy open sleeves
  • 25 Sleeves/Cs.
Specifications
Closure TypeStackable
Bottom ShapeFlat
MaterialPolystyrene
ShapeRound
SterilitySterile
TypeDeep Dish
EdgesRaised Ridge
Packaging16/Sleeve, 25 Sleeves/Cs.
Size100 x 20mm
Height (Metric)20 mm
Diameter (Metric)100 mm
Unit SizeCS