Safety-Labeled Wash Bottles
Safety-Labeled Wash Bottles
Actual product may vary
Safety-Labeled Wash Bottles
Safety-Labeled Wash Bottles
Safety-Labeled Wash Bottles
Fisher Catalogue
Safety-Labeled Wash Bottles
Includes Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols. Fisherbrand™ Safety-Labeled Wash Bottles also include Chemical Abstract Service (CAS) number, protective equipment symbols and target organ inf.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberChemical Label
FISFB0340923MDeionized Water
FISFB0340923KDistilled Water
Catalog number FISFB0340923M
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Chemical Label:
Deionized Water
  • Easy-to-read printed bottles
  • Wide mouth 53mm (2 in.) bottle for easy refilling
  • Translucent, LDPE solvent-resistant; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm; Toluene is red LDPE bottle
  • Polypropylene closures
  • Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
Use only with solvents compatible with LDPE.
Specifications
Capacity (Metric)500 mL
Capacity (English)17 oz.
ColorNatural
MaterialPolyethylene
Closure ColorBlue
Certifications/ComplianceOSHA 29 CFR 1910.1200
AutoclavableNot Autoclavable
MouthWide
Chemical LabelDeionized Water
Closure MaterialPolypropylene
Closure Size53 mm
Unit SizePK