Amber Wide Mouth Packer Glass Bottles with White Polypropylene Caps
Amber Wide Mouth Packer Glass Bottles with White Polypropylene Caps
Actual product may vary
Fisherbrand
Amber Wide Mouth Packer Glass Bottles with White Polypropylene Caps
Fisherbrand™ Amber Wide Mouth Packer Glass Bottles with White Polypropylene Caps are ideal for storage of light sensitive samples or dry powders.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Quantity
FISFB02911452A950 mL36
FISFB02911450A250 mL84
FISFB02911451A500 mL60
Catalog number FISFB02911452A
Price (SGD)
247.00
CS
Add to cart
Capacity (Metric):
950 mL
Quantity:
36
Price (SGD)
247.00
CS
Add to cart
  • Feature a wide opening for making them the ideal general purpose bottle for solids
  • The amber color filters out ultraviolet wavelengths, making amber packers ideal for light sensitive solids
  • Amber packers are the classic vitamin bottles and are often used in pharmaceutical packaging
  • Easy transfer of contents
  • Good impact strength and pliability
  • Caps can be remelted after they are formed
  • Liner is made of a vinyl coating applied to high density polyethylene coated paper and then it is laminated to pulpboard
Specifications
Capacity (Metric)950 mL
ColorAmber
Diameter (Metric) Outer Neck50 mm
Diameter (Metric) Inner Neck40 mm
Height (Metric) With Closure181 mm
MaterialGlass (Type 3)
Quantity36
Closure ColorWhite
Neck Finish53-400
Closure IncludedYes
MouthWide
Height (Metric)178 mm
Product TypeWide Mouth Packer Bottle
Closure LinerPulp/Vinyl
Closure MaterialPolypropylene
Closure Size53-400 mm
Diameter (Metric)99 mm
Unit SizeCS