Product Image
Horizontal Large Gel Systems: Gels
For use with large gel systems
Have Questions?
Catalog NumberFor Use With (Equipment)
FISFB-SB-2530Owl A2 and A5 Large Gel Electrophoresis System
Catalog number FISFB-SB-2530
Price (SGD)
886.00
Each
For Use With (Equipment):
Owl A2 and A5 Large Gel Electrophoresis System
Price (SGD)
886.00
Each
UV-transparent; allows multiple gels to be poured.
Specifications
ColorUV Transparent
For Use With (Equipment)Owl A2 and A5 Large Gel Electrophoresis System
TypeGel Tray
IncludesGasketed end gate
Dimensions (L x W)9.8 x 7.8 in. (25 x 20 cm)
Length (English)9.8 in.
Length (English) Footprint14.75 in.
Length (Metric)25 cm
Length (Metric) Footprint37.5 cm
Width (English)7.8 in.
Width (English) Footprint11 in.
Width (Metric)20 cm
Width (Metric) Footprint28 cm
Height (English) Footprint4.31 in.
Height (Metric) Footprint11 cm
Unit SizeEach