Search Fisher Scientific
| Catalog Number | Capacity (Metric) |
|---|---|
| FISFB-800-1000 | 1000 mL |
| FISFB800100 | 100 mL |
| FKS15446113 | 100 mL |
| FISFB-800-250 | 250 mL |
| FISFB800250 | 250 mL |
| FKS15456113 | 250 mL |
| FISFB-800-500 | 500 mL |
| FISFB800500 | 500 mL |
| FKIFB800-500 | 500 mL |
| FKS15476113 | 500 mL |
| FISFB8001000 | 1000 mL |
| FKS15486113 | 1000 mL |
| FISFB8002000 | 2000 mL |
| FKS15406123 | 2000 mL |
| FISFB8005000 | 5000 mL |
| FKS15416123 | 5000 mL |
| FISFB80010000 | 10,000 mL |
| FKS15436123 | 10,000 mL |