DISTRIMANTM Positive-Displacement Repetitive Pipette
Actual product may vary
Gilson
DISTRIMANTM Positive-Displacement Repetitive Pipette
Positive-displacement, continuously adjustable repeater pipette. DISTRIMANTM Positive-Displacement Repetitive Pipette simplifies multiple dispensing.
Have Questions?
Catalog NumberVolume (Metric)
FISF164001G1 to 1.25 mL
Catalog number FISF164001G
Price (SGD)
2,228.00
Each
Volume (Metric):
1 to 1.25 mL
Price (SGD)
2,228.00
Each
  • Accuracy and Precision: Zero risk of accidental conversion errors with direct read out and exact distribution of volume required by application.
  • Time saving: Save time with continuous adjustable volume and numerous distribution of aliquots: Up to 125. Faster multiple dispensing.
  • Control button for volume selection.
  • DISTRITIPS syringes available in standard and sterilized versions.
  • Only 3 syringes to cover aliquot volumes ranging from 1 μL to 1.25 mL.
Applications:
  • Any application requiring repetitive dispensing in clinical, hospital, biological, chemical, food and beverage, pharmaceutical, cosmetics and forensics fields.
Specifications
Certifications/ComplianceISO-8655
Compatible TipsDistritips
AutoclavableAutoclavable
ColorBlue
MaterialPolypropylene
ModelDISTRIMAN
Number of Channels1
TypePositive Displacement Repetitive Pipette
Volume (Metric)1 to 1.25 mL
For Use With (Application)Viscous and Volatile Solutions
IncludesFunnel Rack, Tubing, Fittings
Quantity1
Warranty1 Year
Length (English)4.25 in.
Width (English)12.48 in.
Height (English)1.42 in.
Fit for PurposeCertified Quality
Unit SizeEach