Check Order Status
RA Lamb Glass Coplin Jar, holds 5 slides (10 back to back)
Actual product may vary
Thermo Scientific
RA Lamb Glass Coplin Jar, holds 5 slides (10 back to back)
Hold up to 10 slides back-to-back with Epredia™ RA Lamb molded glass coplin jars. Jars have a drop-on lid and integral large diameter base.
Have Questions?
Catalog Number
Slide Count
FISE94
10
Catalog number
FISE94
Price (SGD)
-
Slide Count:
10
Features
Heavy, clear glass construction with internal grooves
1.6 in. diameter circular tapered top
Includes drop-on lid
Specifications
Closure Included
Yes
Closure Type
Drop-on lid
Bottom Shape
Round
Material
Glass
Type
Coplin Staining Jar
Slide Count
10
Diameter (English)
1.6 in.