Femtotips™ Microloader Tips for Femtojet Microinjector
Actual product may vary
Fisherbrand
Femtotips™ Microloader Tips for Femtojet Microinjector
Tips to fill microinjection capillaries and recovering surplus solution from capillary
Have Questions?
Catalog NumberFor Use With (Equipment)
FISE5242956003Femtojet microinjector
Catalog number FISE5242956003
Price (SGD)
-
For Use With (Equipment):
Femtojet microinjector
  • With Stringent quality-control
  • Non-filter tip
  • Universal tip fit
  • Volume range: 0.5 - 20μL
  • Color: Light gray
  • Sterilizable rack package
Specifications
For Use With (Application)For recovering surplus solution from the capillary
For Use With (Equipment)Femtojet microinjector
Unit SizePack