Storage Foil Adhesive Seal for Microplates
Storage Foil Adhesive Seal for Microplates
Actual product may vary
Storage Foil Adhesive Seal for Microplates
Storage Foil Adhesive Seal for Microplates
Fisherbrand
Storage Foil Adhesive Seal for Microplates
Storage Film and Foil glued seals for simple and dependable sealing during sample storage
Have Questions?
Catalog NumberMaterial
FISE0030127889Aluminum
Catalog number FISE0030127889
Price (SGD)
709.00
PK
Add to cart
Material:
Aluminum
Price (SGD)
709.00
PK
Add to cart
  • Heat Sealing Film and Foil for continuous locking with the best evaporation protection
  • Foil products are made of aluminum, can be pierced, and protect light-sensitive samples
  • Film products are transparent and provide protection against unwanted punctures
  • Eppendorf Plate Lid: stable, flexible protection of samples during short-term storage
Specifications
MaterialAluminum
Product TypeStorage Foil
Unit SizePK