PCR Cap Strips, Flat
Actual product may vary
Fisherbrand
PCR Cap Strips, Flat
All the benefits of white reflective wells in a PCR-tube format
Have Questions?
Catalog NumberQuantity
FISE0030124847120 Pk.
Catalog number FISE0030124847
Price (SGD)
433.00
Case
Quantity:
120 Pk.
Price (SGD)
433.00
Case
All the benefits of white reflective wells in a PCR-tube format
  • Strips with eight microcaps for 0.1 and 0.2 mL wells
  • Easy and rapid sealing of Eppendorf twin.tec PCR Plates, PCR strips and other standard multiwell plates
  • Flat Cap Strips are suitable for real-time PCR
  • Autoclavable (121 °C, 20 min)
  • Certified free from human DNA, DNase, RNase and PCR inhibitors
Specifications
Certifications/ComplianceHuman DNA, DNase, RNase, PCR inhibitor-free
Quantity120 Pk.
Closure MaterialPolypropylene
For Use With (Equipment)0.1 and 0.2mL tube strips, PCR plates and other standard multiwell plates by providing easy and effective sealing
SterilityNon-sterile
MaterialPolypropylene
Closure TypeFlat Cap
AutoclavableAutoclavable
Format10 x 12 Strips
Product Type8-Strip Flat Cap (no tubes)
Unit SizeCase