Collagen I, Rat
Collagen I, Rat
Actual product may vary
Collagen I, Rat
Enhances cell attachment and proliferation or promotes expression of cell-specific morphology and function. Corning™ Collagen I, Rat is effective as a gel or a thin coating at a recommended concentration of 5 to 10μg/cm2 of growth surface depending on cell type.
Have Questions?
Catalog NumberQuantity
FISCB-40236100 mg
Catalog number FISCB-40236
Price (SGD)
-
Quantity:
100 mg
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Corning offers a broad range of collagen types that are derived from multiple species. These collagens have been used to culture a variety of cell types to promote adhesion, growth, and/or differentiation.
Specifications
Concentration3 to 4 mg/mL
For Use With (Application)Cell Attachment and Proliferation, Cell-specific Morphology and Function
FormulationLiquid in 0.02N acetic acid
Quantity100 mg
SourceRat Tail Tendon
Product TypeCollagen, Rat Tail Type I
Purity or Quality Grade>90% by SDS-PAGE
Unit SizeEA
Contents & Storage
2°C to 8°C