Check Order Status
EZ-Spread™ Plating Beads
Actual product may vary
Genlantis
EZ-Spread™ Plating Beads
Allows you to easily and safely spread bacterial and yeast cells across the surface of an agar plate
Have Questions?
Catalog Number
Quantity
FISC400050
50 Tubes
Catalog number
FISC400050
Price (SGD)
-
Quantity:
50 Tubes
Sterile plating in seconds
No need for glass rods and potentially hazardous ethanol flaming
More uniform spreading and more colonies compared to traditional glass rod method
Two choices of ready-to-use sterile packaging: dispenser bottle for spreading 100 plates or single-use tubes for 50 plating
Fast, easy way to spread cells on multiple plates simultaneously
Safely spread bacterial and yeast cells across the surface of an agar plate
Specifications
For Use With (Application)
Easily and safely spread bacterial and yeast cells across the surface of an agar plate
Quantity
50 Tubes