▸
Recombinant Proteins
▸
Bovine Serum Albumin (BSA) Microbiological Grade Powder
Fisher Bioreagents
Bovine Serum Albumin (BSA) Microbiological Grade Powder
Purified by a modified cold ethanol procedure
Have Questions?
Catalog Number
Quantity
FISBP9700100
100 g
Catalog number
FISBP9700100
Price (SGD)
-
Quantity:
100 g
CAS: 9048-46-8
Specifications
Name
BSA Microbiological Grade
pH Range
6.8 to 7.2
Preparation Method
Modified cold ethanol procedure
Quantity
100 g
Source
Bovine
Form
Powder