Cool Pak™ 2 Microtube and PCR Tube Cooler
Cool Pak™ 2 Microtube and PCR Tube Cooler
Actual product may vary
Cool Pak™ 2 Microtube and PCR Tube Cooler
Cool Pak™ 2 Microtube and PCR Tube Cooler
Cool Pak™ 2 Microtube and PCR Tube Cooler
Cool Pak™ 2 Microtube and PCR Tube Cooler
Protects enzymes and other temperature-sensitive samples from degradation while working on the laboratory bench. Fisher BioReagents™ Cool Pak™ 2 Microtube and PCR Tube Cooler provides temporary cold storage by eliminating the need for messy ice buckets and activates overnight in freezer.
Have Questions?
Catalog NumberdimCapacityMetric
FISBP361036
Catalog number FISBP361036
Price (SGD)
-
  • Ideal for temporarily storing temperature-sensitive samples while setting up reactions
  • Maintains 0°C for 4 hours
  • Dual-sided
  • Holds up to 36 x 1.5/2.0mL microtubes
  • Other side can hold 0.2mL PCR tubes and strips or a 96-well PCR plate with additional capacity for 10 x 1.5/2.0mL microtubes
Specifications
Dimensions (L x W x H)14.3 x 14.3 x 6.4 cm (5.6 x 5.6 x 2.5 in.)
For Use With (Application)Ideal for temporarily storing temperature sensitive samples such as enzymes, Taq, and dNTPs
Weight (English)2 lb.
Weight (Metric)0.9 kg
DescriptionShape: Cube
TypeMicrotube/PCR Tube Cooler
ColorLight Gray
Unit SizeEA