▸
Organic compounds
▸
Organoheterocyclic compounds
▸
Pyridines and derivatives
▸
Pyridoxine Hydrochloride, Fisher BioReagents
Element Search
Structure Search
Pyridoxine Hydrochloride, Fisher BioReagents
Have Questions?
Catalog Number
Quantity
Packaging
FISBP267750
50 g
Amber Glass
Catalog number
FISBP267750
Price (SGD)
-
Quantity:
50 g
Packaging:
Amber Glass
Request bulk or custom format
Specifications
Chemical Name or Material
Pyridoxine Hydrochloride
CAS
58-56-0
ChemAlert Storage Symbol
Gray
Melting Point
214°C
Color
White
View more
Pyridoxine HCl is used as a vitamin (enzyme cofactor).