Check Order Status
TBS, Tris Buffered Saline, 10X Solution, pH 7.4, Molecular Biology, Fisher BioReagents™
Element Search
Structure Search
TBS, Tris Buffered Saline, 10X Solution, pH 7.4, Molecular Biology, Fisher BioReagents™
Have Questions?
Catalog Number
Quantity
Packaging
FISBP2471-100
100 mL
Poly Bottle
Catalog number
FISBP2471-100
Price (SGD)
-
Quantity:
100 mL
Packaging:
Poly Bottle
Request bulk or custom format
Chemical Identifiers
CAS
7447-40-7, 7647-01-0, 7647-14-5, 77-86-1, 7732-18-5
Specifications
Chemical Name or Material
Tris Buffered Saline
Name Note
10X Solution
CAS
7447-40-7
ChemAlert Storage Symbol
Gray
Boiling Point
100°C
View more
TBS, Tris Buffered Saline, 10X Solution, pH 7.4, Molecular Biology
Washing buffer for alkaline phosphatase or peroxidase conjugates in Western blotting