Shaker Flasks
Actual product may vary
Shaker Flasks
Reduce the risk of cross-contamination.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Bottom ShapeClosure Type
FISBBV10001000 mLBaffledVented
FISPBV12-5125 mLPlainVented
FKS11735373125 mLPlainVented
FISBBV12-5125 mLBaffledVented
FISPBV250250 mLPlainVented
FISBBV250250 mLBaffledVented
FISPBV50-0500 mLPlainVented
FISBBV50-0500 mLBaffledVented
FKS11705263500 mLBaffledVented
FISPBV10001000 mLPlainVented
FKS117552531000 mLPlainVented
FKS117752632000 mLPlainVented
FISBBV28002800 mLBaffledVented
Catalog number FISBBV1000
Price (SGD)
312.00
Case
Capacity (Metric):
1000 mL
Bottom Shape:
Baffled
Closure Type:
Vented
Price (SGD)
312.00
Case
Reduce the risk of cross-contamination.
  • Nonpyrogenic and noncytotoxic
  • Clear, durable polycarbonate with molded-in graduations
  • Polypropylene closure with 0.22μm PTFE pore
  • Individually packed for easy storage and handling
  • Plain bottom for use on benchtop or baffled bottom for shaker table use
  • Vented or non-vented screw closure NOT autoclavable
  • Autoclavable flask
  • Sterile
Specifications
Capacity (English)33.8 oz.
Closure MaterialPolypropylene
Capacity (Metric)1000 mL
MaterialPolycarbonate
For Use With (Application)For suspension cell culture, media preparation and storage
Includes0.22μm PTFE pore
AutoclavableAutoclavable
GraduatedYes
SterilitySterile
ClassClass VI
Bottom ShapeBaffled
Closure TypeVented
Unit SizeCase