Drosophila Vials
Actual product may vary
Drosophila Vials
Straight wall design minimizes the dislodging of food; provides more efficient racking, filling and plugging.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric) OuterTypeMaterialMouth Style
FISAS50725.12 mmDrosophila Vial, Bulk PackedPolypropyleneNarrow
FISAS51929.21 mmDrosophila Vial, Bulk PackedPolystyreneWide
FKS1582027529.21 mmDrosophila Vial, Tray PackedPolypropyleneWide
Catalog number FISAS507
Price (SGD)
141.00
Case
Diameter (Metric) Outer:
25.12 mm
Type:
Drosophila Vial, Bulk Packed
Material:
Polypropylene
Mouth Style:
Narrow
Price (SGD)
141.00
Case
Straight wall design minimizes the dislodging of food; provides more efficient racking, filling and plugging
Fisherbrand Drosophila vials are available in clear polystyrene, translucent and flexible polypropylene, and virtually unbreakable K-resin. These vials feature a flat bottom, straight wall design for convenient racking, filling and stacking. Available bulk packaged or in a convenient, 10 x 10 tray pack configuration.
Polypropylene Vials
  • Translucent and flexible
  • Autoclavable
Polystyrene Vials
  • Excellent clarity for optimal visibility
  • Nonautoclavable
K-Resin Vials
  • Highly flexible, virtually unbreakable
  • Nonautoclavable
Standard-Packed Glass Shell Vials
  • Type 1, N-51A borosilicate glass
  • Securely packed in cardboard trays with individual vial dividers
Bulk- and Tray-Packed Shell Vials
  • Available in narrow- or wide-mouth styles
  • Constructed from polystyrene, K-Resin™, or polypropylene
  • K-Resin and polypropylene vials are virtually unbreakable, scratch and mar resistant; polystyrene and K-Resin vials have glass-like clarity
  • Polypropylene vials are autoclavable; polystyrene and K-Resin are nonautoclavable
Specifications
AutoclavableAutoclavable
ColorClear
Diameter (English) Outer0.98 in.
Diameter (Metric) Outer25.12 mm
For Use With (Application)Straight wall design minimizes the dislodging of food, provides more efficient racking, filling and plugging
Height (English)3.74 in.
Height (Metric)95 mm
Mouth StyleNarrow
MaterialPolypropylene
TypeDrosophila Vial, Bulk Packed
Unit SizeCase