Drosophila Supplies Petri Dishes
Drosophila Supplies Petri Dishes
Actual product may vary
Drosophila Supplies Petri Dishes
Ideal for embryo collection and nematode research
Have Questions?
Catalog NumberVent
FISAS4051No
Catalog number FISAS4051
Price (SGD)
391.00
CS
Add to cart
Vent:
No
Price (SGD)
391.00
CS
Add to cart
Ideal for embryo collection and nematode research
  • Use with the AS-225 Bottle Adapter Ring
  • Engineered for optimum flatness and uniform thickness
  • Packaged sterile in easy-to-open bags
  • 20 per sleeve, 500 per case
Specifications
SterilitySterile
PackagingEasy-to-open bags
Quantity20/Sleeve, 500 Case
VentNo
Height (Metric)15 mm
Diameter (Metric)60 mm
Unit SizeCS