▸
Salts and Inorganics
▸
Inorganic Salts
▸
Alkali Metal Salts
▸
Potassium Nitrate, 1 M, Filling Solution, Fisherbrand
Element Search
Structure Search
Potassium Nitrate, 1 M, Filling Solution, Fisherbrand
Have Questions?
Catalog Number
Quantity
FISACCU0834500
500 mL
Catalog number
FISACCU0834500
Price (SGD)
-
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or Material
Potassium Nitrate
Name Note
1M, Fill Solution
CAS
7732-18-5
CAS
7757-79-1
Physical Form
Liquid
View more