▸
Salts and Inorganics
▸
Inorganic Salts
▸
Alkali Metal Salts
▸
Sodium Sulfate, 1 M, Filling Solution, Fisherbrand
Element Search
Structure Search
Sodium Sulfate, 1 M, Filling Solution, Fisherbrand
Have Questions?
Catalog Number
Quantity
FISACCU0833500
500 mL
Catalog number
FISACCU0833500
Price (SGD)
-
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or Material
Sodium Sulfate
Name Note
1M, Fill Solution
CAS
7732-18-5
CAS
7757-82-6
Physical Form
Liquid
View more