Domed and Flat PCR Cap Strips
Domed and Flat PCR Cap Strips
Actual product may vary
Fisherbrand
Domed and Flat PCR Cap Strips
Optimize PCR applications with these cap strips for use with Thermo Scientific 96-well PCR plates and strip tubes.
Have Questions?
Catalog NumberQuantityColor
FISAB-078625 packs of 12Natural
Catalog number FISAB-0786
Price (SGD)
340.00
CS
Add to cart
Quantity:
25 packs of 12
Color:
Natural
Price (SGD)
340.00
CS
Add to cart
These PCR Cap Strips are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.
Highlights
  • Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
  • Large end tabs for strip removal (12 strips only)
Specifications
ColorNatural
Certifications/ComplianceCertified free of human DNA, RNase, DNase, and endotoxins.
MaterialVirgin Medical Grade Polypropylene
No. of Wells96
No. per Case300
No. per Pack12
Quantity25 packs of 12
For Use With (Application)Facilitates sample labeling
FormatStrips of 8 Flat Caps
Plate Compatibility96-well PCR Plate, Standard and Low-profile PCR Tube Strips
Unit SizeCS