MOPS, ≥98.5% (Dry Basis), Ultrapure
MOPS, ≥98.5% (Dry Basis), Ultrapure
MOPS, ≥98.5% (Dry Basis), Ultrapure
MOPS, ≥98.5% (Dry Basis), Ultrapure
Thermo Scientific
MOPS, ≥98.5% (Dry Basis), Ultrapure
CAS: 1132-61-2 | C7H15NO4S | 209.26 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FISAAJ19256A11 kgPlastic bottle
Catalog number FISAAJ19256A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialMOPS
CAS1132-61-2
Melting Point277°C to 280°C (decomposition)
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c