Check Order Status
Formic Acid, 88% (Certified ACS), Fisher Chemical™
Element Search
Structure Search
Formic Acid, 88% (Certified ACS), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FISA118P-500
500 mL
Poly Bottle
Catalog number
FISA118P-500
Price (SGD)
-
Quantity:
500 mL
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Formic Acid, 88%
Cleanup Symbol
Acid
pH
2.1
CAS
64-18-6
ChemAlert Storage Symbol