Isotemp™ Undercounter BOD Refrigerated Incubator
Isotemp™ Undercounter BOD Refrigerated Incubator
Actual product may vary
Isotemp™ Undercounter BOD Refrigerated Incubator
Fisher Scientific Isotemp low temperature incubators are an ideal choice for incubation above or below ambient temperature.
Have Questions?
Catalog NumberCapacity (Metric)
FIS97-990E172.2 L
Catalog number FIS97-990E
Price (SGD)
15,892.00
EA
Add to cart
Capacity (Metric):
172.2 L
Price (SGD)
15,892.00
EA
Add to cart
Fisher Scientific Isotemp low temperature incubators are an ideal choice for incubation above or below ambient temperature.  Low temperature, refrigerated incubators are suited to fit your application needs for BOD (biochemical oxygen demand) waste water testing, food, beverage and agricultural testing, pharmaceutical research, microbiology, yeast growth and more.
Key features:
  • Temperature specifications cover BOD test standards
  • Outstanding temperature uniformity and stability
  • Temperature alarm designed to protect your samples
  • Wide temperature range from as low as -10°C up to 60°C*
  • Easy-to-use microprocessor control
  • Easy-to-clean, corrosion-resistant construction
  • Cooling system designed for performance and long-life**
  • 2-year warranty on parts and labor
  • cCSAus listed
Under-counter model (173 L/  6.1 cu. ft.) for smaller sample volume
  • Space-saving design
  • Run electrical devices inside incubator with interior outlet (2A)
  • Holds up to 114 BOD bottles
  • LED display with read-out to 0.1°C
Note: Free-Standing model (566L, 20 cu. ft.) is also available.  See Cat. No. 3720A.
*Refer to specifications for additional information.
**Based on internal testing
Specifications
Capacity (English)6.1 cu. ft.
Certifications/ComplianceCSA
Capacity (Metric)172.2 L
Chamber MaterialPlastic
Temperature Range (Metric)-10°C to +60°C
TypeRefrigerated Incubator
DisplayLED
Door StyleDoor is not lockable.
Hood MaterialEnamel Steel
IncludesTwo removable shelves, three molded door shelves, line cord and plug
InsulationFoamed Polyurethane
ModelUnder-counter model
Temperature Uniformity±1°C
Dimensions (L x W x H)24.5 x 24 x 34.5 in. (62.2 x 60.96 x 87.6 cm)
Length (English)24.5 in.
Length (Metric)62.2 cm
Width (English)24 in.
Width (Metric)60.96 cm
Height (English)34.5 in.
Height (Metric)87.6 cm
Electrical Requirements115 V, 60 Hz
Voltage115 V
Amperage6 A
Frequency60 Hz
Unit SizeEA