▸
Buffers and Standards
▸
Matrix Blanks and Standardization Solutions
▸
Zero Oxygen Standard, for Calibration of Dissolved Oxygen Meters, Ricca Chemical
Element Search
Structure Search
Zero Oxygen Standard, for Calibration of Dissolved Oxygen Meters, Ricca Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FIS94204
120 mL
Natural Poly Bottle
Catalog number
FIS94204
Price (SGD)
-
Quantity:
120 mL
Packaging:
Natural Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Zero Oxygen Standard
Name Note
For Calibration of Dissolved Oxygen Meters
Packaging
Natural Poly Bottle
CAS
7791-13-1
CAS Min %
93.0
View more