3DHistech SlideCenter Slide Management System - 5 Users
Actual product may vary
3DHistech SlideCenter Slide Management System - 5 Users
Epredia™ 3DHistech SlideCenter is a powerful, server-based slide management system with a fully featured slide database capable of storing both macroscopic images and digital slides. It can be adapted to several fields, including various research applications and medical education.
Have Questions?
Catalog number FIS9190-1
Price (SGD)
-
SlideCenter is a flexible, distributed server system: more servers can easily be added for enhanced data security and storage requirements.
Licensing is performed on the server side: SlideCenter regulates the number of users who can access the server at any time.
Five user types can be defined: Administrator user, Moderator user, Normal user, Restricted user and Public Share user.
Key Features
  • Automatic slide alignment
  • Virtual Slide and VirtualTray
  • TMA workflow support
  • Transparent login method for high-traffic user authentication
  • Server-side barcode parsing
  • InstantViewer
  • Web-based slide and case database with fast search
  • Easy extension by adding new SUs
  • Slide access through the free SlideViewer, the InstantViewer or the iPad Viewer
  • Virtual Slide handling
  • MS Network, HTTP and HTTPS accessibility
  • Fully manageable user and user group rights system
  • Direct scanning into database with PANNORAMIC digital slide scanners
  • Macro images and multiple file attachments stored in the case
  • SlideCenter can work with other 3DHISTECH software applications (e.g. quantification software can be run on remote slides)
  • License for 5 users - Research use only.
For research and education use only, not for use in diagnostic procedures in the U.S. This product has not been approved or cleared as a medical device by the U.S. Food and Drug Administration.
Specifications
Product TypeSlide Management System
For Use With (Application)Digital Pathology
For Use With (Equipment)Microarray Scanner
Unit SizeEach