Turbidity Standard, Primary Standard Stock Formazin Suspension, 4000 NTU, Ricca Chemical
Turbidity Standard, Primary Standard Stock Formazin Suspension, 4000 NTU, Ricca Chemical
Actual product may vary
Turbidity Standard, Primary Standard Stock Formazin Suspension, 4000 NTU, Ricca Chemical
Turbidity Standard, Primary Standard Stock Formazin Suspension, 4000 NTU, Ricca Chemical
Turbidity Standard, Primary Standard Stock Formazin Suspension, 4000 NTU, Ricca Chemical
Turbidity Standard, Primary Standard Stock Formazin Suspension, 4000 NTU packaged in an amber glass bottle.
Have Questions?
Catalog NumberVolume (Metric)
FIS8830-16500 mL
Catalog number FIS8830-16
Price (SGD)
-
Volume (Metric):
500 mL
Turbidity Standard; Primary Standard Stock Formazin Suspension, 4000 NTU (Nephelometric Turbidity Units); Ricca Chemical; 500 mL; Amber glass
  • Turbidity Standard
  • Primary Standard Stock Formazin Suspension, 4000 NTU (Nephelometric Turbidity Units)
  • Amber glass
Specifications
TypeTurbidity Standard
For Use With (Equipment)Turbidimeter
DescriptionPrimary Standard Stock Formazin Suspension, 4000 NTU (Nephelometric Turbidity Units)
Volume (English)16.9 oz.
Volume (Metric)500 mL
Packaging TypeAmber Glass
Shelf Life12 Months
Unit SizeEA