Check Order Status
Sulfuric Acid, 0.250 N (N/4), Ricca Chemical
Element Search
Structure Search
Sulfuric Acid, 0.250 N (N/4), Ricca Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FIS8270-32
1 L
Natural Poly Bottle
Catalog number
FIS8270-32
Price (SGD)
-
Quantity:
1 L
Packaging:
Natural Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Sulfuric Acid
Color
Colorless
CAS
7732-18-5
CAS Min %
97.0
Concentration
0.250N
View more