▸
Facility Safety and Maintenance
▸
Maintenance Lubricants and Penetrating Fluids
▸
Apiezon Grease
Apiezon Grease
Lubricate pumps with this grease. Fisherbrand™ Apiezon Grease is designed for use with Fisherbrand™ rotary vane pumps.
Have Questions?
Catalog Number
Quantity
Product Type
FIS8000-16
50 g
L Grease
Catalog number
FIS8000-16
Price (SGD)
-
Quantity:
50 g
Product Type:
L Grease
Apiezon grease
For general use in Fisherbrand rotary vane pumps
Specifications
Product Type
L Grease
Pressure
20°C 7x10
-11
torr
Quantity
50 g
Temperature (Metric)
10°C to 30°C