▸
Salts and Inorganics
▸
Inorganic Salts
▸
Alkali Metal Salts
▸
Sodium Chloride, 1.00 M, Ricca Chemical
Element Search
Structure Search
Fisherbrand
Sodium Chloride, 1.00 M, Ricca Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FIS7227-132
1 L
Natural Poly Bottle
Catalog number
FIS7227-132
Price (SGD)
-
Quantity:
1 L
Packaging:
Natural Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Sodium Chloride
Shelf Life
24 months
CAS
7732-18-5
Color
Colorless
CAS Min %
93.0
View more