1x1 image pixel for data collection
Ammonium Carbonate Solution R, 158 g/L, Ricca Chemical
Ammonium Carbonate Solution R, 158 g/L, Ricca Chemical
Ammonium Carbonate Solution R, 158 g/L, Ricca Chemical
Ammonium Carbonate Solution R, 158 g/L, Ricca Chemical
Ammonium Carbonate Solution R, 158 g/L, Ricca Chemical
Ammonium Carbonate Solution R, 158 g/L, Ricca Chemical
Have Questions?
Catalog NumberQuantity
FIS62215-16500 mL
Catalog number FIS62215-16
Price (SGD)
-
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium Carbonate Solution
ColorColorless
Name NoteReagent
CAS506-87-6
CAS Min %85.0
View more
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link