Check Order Status
Potassium Chloride Reference Solution C, 1408.8 μS/cm at 25°C, 0.01 M, Ricca Chemical
Element Search
Structure Search
Potassium Chloride Reference Solution C, 1408.8 μS/cm at 25°C, 0.01 M, Ricca Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FIS5895-32
1 L
Natural Poly Bottle
Catalog number
FIS5895-32
Price (SGD)
-
Quantity:
1 L
Packaging:
Natural Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Potassium Chloride Reference Solution C
Name Note
1408.8μS/cm (1408.8μmho/cm) at 25°C
Packaging
Natural Poly Bottle
CAS
7447-40-7
CAS Min %
98.0
View more