Check Order Status
Potassium Chloride Conductivity Standard, 10,000 μS/cm (10 mS/cm) at 25°C, Ricca Chemical
Element Search
Structure Search
Potassium Chloride Conductivity Standard, 10,000 μS/cm (10 mS/cm) at 25°C, Ricca Chemical
Have Questions?
Catalog Number
Quantity
FIS588810-16
500 mL
Catalog number
FIS588810-16
Price (SGD)
-
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or Material
Potassium Chloride Conductivity Standard
Name Note
10 mS/cm (10, 000μmho/cm) at 25°C
Packaging
Natural Poly Bottle
CAS
7447-40-7
CAS Min %
97.0
View more