Check Order Status
Potassium Chloride Conductivity Standard, 1000 μS/cm at 25°C, Ricca Chemical
Element Search
Structure Search
Potassium Chloride Conductivity Standard, 1000 μS/cm at 25°C, Ricca Chemical
Tested in ISO 17025 accredited laboratories, providing the utmost quality in calibration.
Have Questions?
Catalog Number
Quantity
Packaging
FIS588801-16
500 mL
Natural Poly Bottle
Catalog number
FIS588801-16
Price (SGD)
-
Quantity:
500 mL
Packaging:
Natural Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Potassium Chloride Conductivity Standard
Name Note
1 mS/cm (1000μmho/cm) at 25°C
Packaging
Natural Poly Bottle
CAS
7447-40-7
CAS Min %
98.0%
View more
Tested in ISO 17025 Accredited Laboratories.
Manufactured in cGMP compliant, FDA registered facilities.
Certified Traceable to NIST Standard Reference Material when available.
Many C of As and labels contain TDS and conductance information.
Tightest Specifications in the industry, lowest lot-to-lot variability.