Check Order Status
Potassium Bromate RV, EP Primary Standard, Ricca Chemical
Element Search
Structure Search
Potassium Bromate RV, EP Primary Standard, Ricca Chemical
Have Questions?
Catalog Number
Quantity
FIS58794-500
500 g
Catalog number
FIS58794-500
Price (SGD)
-
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or Material
Potassium Bromate RV
Shelf Life
24 months
Name Note
Primary Standard
CAS
7732-18-5
Color
White
View more