Check Order Status
ORP Standard, +600 mV, Ricca Chemical
Element Search
Structure Search
ORP Standard, +600 mV, Ricca Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FIS54646-16
500 mL
Amber Glass Bottle
Catalog number
FIS54646-16
Price (SGD)
-
Quantity:
500 mL
Packaging:
Amber Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Oxidation-Reduction Potential Standard
Shelf Life
9 months
Name Note
+600mV vs. Ag/AgCl
Packaging
Amber Glass Bottle
CAS
7783-85-9
View more