Check Order Status
Fluoride Standard, 2 ppm F
-
(Premixed with TISAB/Color Coded Red), Ricca Chemical
Element Search
Structure Search
Fluoride Standard, 2 ppm F
-
(Premixed with TISAB/Color Coded Red), Ricca Chemical
Have Questions?
Catalog Number
Quantity
FIS3163-16
500 mL
Catalog number
FIS3163-16
Price (SGD)
-
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS
7681-49-4, 1310-73-2, 64-19-7, 7647-14-5, 13291-61-7, 25956-17-6, 7732-18-5
Specifications
Chemical Name or Material
Fluoride Standard
Packaging
Natural Poly Bottle
CAS
7681-49-4
CAS Min %
93.0
Concentration or Composition Notes
2ppm F