Check Order Status
HP35 Disposable Microtome Blades
Actual product may vary
Thermo Scientific
HP35 Disposable Microtome Blades
Epredia™ HP35 microtome blades are coated for reliable and thin sectioning.
Have Questions?
Catalog Number
Coating
FIS3150734
Coated
Catalog number
FIS3150734
Price (SGD)
487.00
CS
Add to cart
Coating:
Coated
Price (SGD)
487.00
CS
Add to cart
High-profile
Increased durability
Specifications
Product Type
High Profile Blade
Profile
High
For Use With (Application)
Designed for high-quality thin sectioning
Disposable
Single-use
Coating
Coated
Cutting Angle
35°
Thickness (English)
0.01 in.
Thickness (Metric)
0.31 mm