Glass Thermometer
Actual product may vary
Fisherbrand
Glass Thermometer
Glass Thermometer features a 10/18 joint and a kerosene bulb.
Have Questions?
Catalog NumberTemperature Range (Metric)Immersion Length
FIS31-502-350-20°C to +250°C25 mm
Catalog number FIS31-502-350
Price (SGD)
163.00
Each
Temperature Range (Metric):
-20°C to +250°C
Immersion Length:
25 mm
Price (SGD)
163.00
Each
  • Ground glass thermometer.
  • Immersion length is measured from the bottom of the joint to the lower end of the kerosene bulb.
  • Accuracy: ±1°C.
  • 10/18 joint.
Specifications
For Use With (Application)Temperature Monitoring
Immersion TypePartial Immersion
Length (Metric)305 mm
MaterialBorosilicate Glass
Thermometer FillKerosene
Product TypeThermometer
Description10/18 Joint
Max. Temperature (English)482°F
Max. Temperature (Metric)250°C
Min. Temperature (English)-4°F
Min. Temperature (Metric)-20°C
Temperature Range (English)-4°F to +482°F
Temperature Range (Metric)-20°C to +250°C
For Use With (Equipment)Ground Glass Thermometer Adapter
Accuracy±1°C
Subdivision20°C
Immersion Length25 mm
Unit SizeEach