Schlenk Flasks, Modified, with Glass Stopcock
Actual product may vary
Fisherbrand
Schlenk Flasks, Modified, with Glass Stopcock
Kjeldahl shape to facilitate heating and stirring
Have Questions?
Catalog NumberCapacity (Metric)Standard Taper Joint
FIS31-501-35510 mL14/20
Catalog number FIS31-501-355
Price (SGD)
158.00
Each
Capacity (Metric):
10 mL
Standard Taper Joint:
14/20
Price (SGD)
158.00
Each
  • Provides easy transfer of liquids and solids
  • Glass stopcock with 2 mm bore
  • Top with standard taper outer joint
Specifications
DescriptionSchlenk Flask, Modified
Capacity (Metric)10 mL
MaterialBorosilicate Glass (3.3)
Standard Taper Joint14/20
AutoclavableAutoclavable
DisposableReusable
GraduatedNo
Mouth StyleRimmed
SterilityNon-sterile
ClassA
Bottom ShapeRound
Closure TypeStopper
Volume (Metric) Working10 mL
Closure Size14/20
Unit SizeEach