Liebig Condenser
Actual product may vary
Synthware
Liebig Condenser
With standard taper outer joint at the top and lower inner drip tip joint
Have Questions?
Catalog NumberStandard Taper Joint NoLength (Metric) Jacket
FIS31-500-81014/20120 mm
Catalog number FIS31-500-810
Price (SGD)
215.00
Each
Standard Taper Joint No:
14/20
Length (Metric) Jacket:
120 mm
Price (SGD)
215.00
Each
  • Joints of same size
Specifications
DescriptionHose Connection: 8mm O.D.
For Use With (Application)Distillation
For Use With (Equipment)Distillation Head
IncludesRC14, R201402
Length (Metric) Jacket120 mm
No. of BulbsNone
Standard Taper Joint No14/20
MaterialGlass
TypeLiebig Condenser
Unit SizeEach