Check Order Status
MX35 Premier™ Disposable Low-profile Microtome Blades
Actual product may vary
Fisherbrand
MX35 Premier™ Disposable Low-profile Microtome Blades
Section hard tissues and high-quality thin sections with high-performance with Epredia™ MX35 Premier™ disposable low-profile microtome blades.
Have Questions?
Change view
Catalog Number
Product Type
FIS3052835
Microtome Blade, Premier Plus
FIS3051835
Microtome Blade, Premier
2 Options
Catalog number
FIS3052835
Price (SGD)
446.00
CS
Add to cart
Product Type:
Microtome Blade, Premier Plus
Price (SGD)
446.00
CS
Add to cart
Sharper edge with increased durability for longer blade life
Premier Plus blades provide greater durability, superior sharpness for lower blade friction and longer blade life
Specifications
Product Type
Microtome Blade, Premier Plus
Profile
Low
For Use With (Application)
Designed to be extra sharp with an extended cutting life (excellent for hard tissues)
Disposable
Single-use
Cutting Angle
34°
Thickness (English)
0.01 in.
Thickness (Metric)
0.24 mm