▸
Histology and Cytology
▸
Microtomes and Accessories
▸
MB35 Premier Disposable Microtome Blade
Actual product may vary
Thermo Scientific
MB35 Premier Disposable Microtome Blade
Use the Epredia™ MB35 premier disposable microtome blade for routine sectioning applications.
Have Questions?
Catalog Number
Profile
FIS3050835
Low
Catalog number
FIS3050835
Price (SGD)
-
Profile:
Low
Low-profile
Increased durability
Specifications
Product Type
Low Profile Blade
Profile
Low
For Use With (Application)
Best for routine sectioning applications
Disposable
Single-use
Cutting Angle
35°
Thickness (English)
0.01 in.
Thickness (Metric)
0.25 mm