Fehling's Copper Solution (Fehling Solution A), Ricca Chemical
Fehling's Copper Solution (Fehling Solution A), Ricca Chemical
Fehling's Copper Solution (Fehling Solution A), Ricca Chemical
Fehling's Copper Solution (Fehling Solution A), Ricca Chemical
Fehling's Copper Solution (Fehling Solution A), Ricca Chemical
Fehling's Copper Solution (Fehling Solution A), Ricca Chemical
For Determination of Reducing Sugars
Have Questions?
Catalog NumberQuantityPackaging
FIS3010-16500 mLNatural Poly Bottle
Catalog number FIS3010-16
Price (SGD)
-
Quantity:
500 mL
Packaging:
Natural Poly Bottle
Request bulk or custom format
Chemical Identifiers
CAS7758-99-8, 7732-18-5
Specifications
Chemical Name or MaterialFehling's Copper Solution
ColorBlue
Name NoteFehling Solution A
CAS7758-99-8
CAS7732-18-5
View more