Check Order Status
Diphenylamine Solution R, Ricca Chemical
Element Search
Structure Search
Diphenylamine Solution R, Ricca Chemical
EP Reagents
Have Questions?
Catalog Number
Quantity
FIS25-834
120 mL
Catalog number
FIS25-834
Price (SGD)
-
Quantity:
120 mL
Request bulk or custom format
Specifications
Chemical Name or Material
Diphenylamine Solution R
Shelf Life
12 months
Color
Colorless
CAS
7664-93-9
CAS Min %
97.95
View more