Amber Wide Mouth Packer Bottles with White Polypropylene PE Foam Cap
Amber Wide Mouth Packer Bottles with White Polypropylene PE Foam Cap
Actual product may vary
Qorpak
Amber Wide Mouth Packer Bottles with White Polypropylene PE Foam Cap
General use bottles ideal for UV sensitive contents. Qorpak™ Amber Wide Mouth Packer Bottles with White Polypropylene PE Foam Cap are used in chemical and environmental applications.
Have Questions?
Catalog NumberCapacity (Metric)Quantity
FIS239213250 mL24/Cs.
Catalog number FIS239213
Price (SGD)
236.00
CS
Add to cart
Capacity (Metric):
250 mL
Quantity:
24/Cs.
Price (SGD)
236.00
CS
Add to cart
  • Feature a wide opening for making them the ideal general purpose bottle for solids
  • Classic "vitamin bottles" and are often used in pharmaceutical packaging
Specifications
Capacity8.5 oz., 250mL
Capacity (Metric)250 mL
Capacity (English)8.5 oz.
ColorAmber
Diameter (English)2.56 in.
MaterialGlass
Quantity24/Cs.
Closure ColorWhite
Height (Metric)119 mm
Height (English)4.69 in.
Closure LinerPTFE
Closure MaterialPolypropylene
Closure Size45-400 mm
Diameter (Metric)65 mm
Closure TypeScrew Cap
Unit SizeCS